siRNA Details
| Gene Symbol: Acap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207223.1 | NP_997106.1 | 1, 2 | 575 | ||
| GenBank mRNA | AK147505.1 | - | 1, 2 | 629 | ||
| BC025556.1 | - | 1, 2 | 149 | |||
| BC067016.1 | - | 1, 2 | 575 | 6827624 | ||
| BC098196.1 | - | 1, 2 | 151 | 6847993 | ||

Acap3
Genomic Map