siRNA Details
| Gene Symbol: Acap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207223.1 | NP_997106.1 | 22 | 2697 | ||
| GenBank mRNA | AK044515.1 | - | 3 | 323 | ||
| AK145262.1 | - | 16 | 1665 | |||
| AK147505.1 | - | 22 | 2751 | |||
| AK153961.1 | - | 3 | 378 | |||
| AK159055.1 | - | 18 | 2567 | |||
| BC025556.1 | - | 21 | 2521 | |||
| BC026407.1 | - | 4 | 414 | |||
| BC067016.1 | - | 22 | 2697 | 6827624 | ||
| BC098196.1 | - | 21 | 2510 | 6847993 | ||

Acap3
Genomic Map