siRNA Details
| Gene Symbol: Gpi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207592.1 | NP_997475.1 | 18 | 1730 | ||
| GenBank mRNA | BC062005.1 | - | 18 | 1730 | 5599776 | |
| FQ215247.1 | - | 7 | 777 | |||
| FQ216077.1 | - | 7 | 778 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpi| Gene Symbol: Gpi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207592.1 | NP_997475.1 | 18 | 1730 | ||
| GenBank mRNA | BC062005.1 | - | 18 | 1730 | 5599776 | |
| FQ215247.1 | - | 7 | 777 | |||
| FQ216077.1 | - | 7 | 778 | |||