siRNA Details
Gene Symbol: Gpi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207592.1 | NP_997475.1 | 6 | 582 | ||
GenBank mRNA | BC062005.1 | - | 6 | 582 | 5599776 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gpi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207592.1 | NP_997475.1 | 6 | 582 | ||
GenBank mRNA | BC062005.1 | - | 6 | 582 | 5599776 |