siRNA Details
| Gene Symbol: Rtcb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207614.1 | NP_997497.1 | 7 | 704 | ||
| GenBank mRNA | AY572415.1 | - | 7 | 704 | ||
| BC078924.1 | - | 7 | 757 | 7106266 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rtcb| Gene Symbol: Rtcb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207614.1 | NP_997497.1 | 7 | 704 | ||
| GenBank mRNA | AY572415.1 | - | 7 | 704 | ||
| BC078924.1 | - | 7 | 757 | 7106266 | ||