siRNA Details
| Gene Symbol: Yipf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207633.2 | NP_997516.1 | 2, 3 | 641 | ||
| GenBank mRNA | AK020780.1 | - | 2, 3 | 215 | ||
| AK030473.1 | - | 2, 3 | 643 | |||
| AK045453.1 | - | 2, 3 | 353 | |||
| AK053562.1 | - | 2, 3 | 642 | |||
| AK079683.1 | - | 2, 3 | 253 | |||
| AK143152.1 | - | 2, 3 | 211 | |||
| AK160251.1 | - | 3, 4 | 269 | |||
| BC048712.1 | - | 2, 3 | 276 | 6328657 | ||

Yipf6
Genomic Map