siRNA Details
| Gene Symbol: AU019823 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134902.1 | NP_001128374.1 | 7 | 773 | ||
| NM_212449.2 | NP_997614.2 | 6 | 746 | |||
| GenBank mRNA | AK133777.1 | - | 5 | 2556 | ||
| BC067205.1 | - | 6 | 528 | 6335744 | ||
| BY740784.1 | - | 7 | 590 | |||

AU019823
Genomic Map