siRNA Details
| Gene Symbol: Cfb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212466.3 | NP_997631.2 | 10 | 1563 | ||
| GenBank mRNA | BC087089.1 | - | 10 | 1563 | 7190440 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cfb| Gene Symbol: Cfb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212466.3 | NP_997631.2 | 10 | 1563 | ||
| GenBank mRNA | BC087089.1 | - | 10 | 1563 | 7190440 | |