siRNA Details
| Gene Symbol: Atp6v1g2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212490.2 | NP_997655.1 | 3 | 1305 | ||
| XM_008772735.1 | XP_008770957.1 | 3 | 1096 | |||
| XM_008772736.1 | XP_008770958.1 | 3 | 1076 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atp6v1g2| Gene Symbol: Atp6v1g2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212490.2 | NP_997655.1 | 3 | 1305 | ||
| XM_008772735.1 | XP_008770957.1 | 3 | 1096 | |||
| XM_008772736.1 | XP_008770958.1 | 3 | 1076 | |||