siRNA Details
| Gene Symbol: Cuta | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164706.1 | NP_001158178.1 | 6 | 462 | ||
| NM_001164707.1 | NP_001158179.1 | 5 | 515 | |||
| NM_212494.2 | NP_997659.2 | 6 | 515 | |||
| XM_006256120.2 | XP_006256182.1 | 7 | 546 | |||
| GenBank mRNA | BP501235.1 | - | 5 | 529 | ||
| CB586341.1 | - | 6 | 462 | |||
| DV717217.1 | - | 5 | 487 | |||
| FQ220708.1 | - | 6 | 749 | |||
| FQ223106.1 | - | 4 | 189 | |||
| FQ229047.1 | - | 4 | 189 | |||

Cuta
Genomic Map