siRNA Details
| Gene Symbol: Dxo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212497.2 | NP_997662.1 | 4 | 811 | ||
| GenBank mRNA | BC082083.1 | - | 4 | 811 | 7133445 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dxo| Gene Symbol: Dxo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212497.2 | NP_997662.1 | 4 | 811 | ||
| GenBank mRNA | BC082083.1 | - | 4 | 811 | 7133445 | |