siRNA Details
| Gene Symbol: Nrm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212508.2 | NP_997673.1 | 4 | 773 | ||
| XM_008772732.1 | XP_008770954.1 | 4 | 1896 | |||
| GenBank mRNA | BC098637.1 | - | 4 | 925 | ||
| FQ220138.1 | - | 3 | 554 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nrm| Gene Symbol: Nrm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212508.2 | NP_997673.1 | 4 | 773 | ||
| XM_008772732.1 | XP_008770954.1 | 4 | 1896 | |||
| GenBank mRNA | BC098637.1 | - | 4 | 925 | ||
| FQ220138.1 | - | 3 | 554 | |||