siRNA Details
| Gene Symbol: Atp5l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212516.2 | NP_997681.1 | 3 | 351 | ||
| GenBank mRNA | BC058497.1 | - | 3 | 352 | 6918545 | |
| FQ210248.1 | - | 3 | 341 | |||
| FQ211867.1 | - | 3 | 348 | |||
| FQ214576.1 | - | 347 | ||||
| FQ217032.1 | - | 3 | 334 | |||
| FQ217139.1 | - | 3 | 367 | |||
| FQ217410.1 | - | 3 | 368 | |||
| FQ218008.1 | - | 3 | 340 | |||
| FQ223878.1 | - | 3 | 341 | |||
| FQ223920.1 | - | 3 | 347 | |||
| FQ224004.1 | - | 3 | 341 | |||
| FQ224067.1 | - | 3 | 341 | |||
| FQ224644.1 | - | 3 | 347 | |||
| FQ229120.1 | - | 3 | 382 | |||

Atp5l
Genomic Map