siRNA Details
| Gene Symbol: Tomm40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212520.1 | NP_997685.1 | 1 | 17 | ||
| XM_006228420.2 | XP_006228482.1 | 2 | 125 | |||
| GenBank mRNA | AB162855.1 | - | 1 | 17 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tomm40| Gene Symbol: Tomm40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212520.1 | NP_997685.1 | 1 | 17 | ||
| XM_006228420.2 | XP_006228482.1 | 2 | 125 | |||
| GenBank mRNA | AB162855.1 | - | 1 | 17 | ||