siRNA Details
| Gene Symbol: Tomm40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212520.1 | NP_997685.1 | 7 | 813 | ||
| GenBank mRNA | AB162855.1 | - | 7 | 813 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tomm40| Gene Symbol: Tomm40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212520.1 | NP_997685.1 | 7 | 813 | ||
| GenBank mRNA | AB162855.1 | - | 7 | 813 | ||