siRNA Details
| Gene Symbol: Tyrobp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212525.1 | NP_997690.1 | 4, 5 | 288 | ||
| XM_008759166.1 | XP_008757388.1 | 4, 5 | 348 | |||
| GenBank mRNA | AY247021.1 | - | 4, 5 | 288 | ||
| FQ211365.1 | - | 4, 5 | 294 | |||

Tyrobp
Genomic Map