siRNA Details
| Gene Symbol: Kcne4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212526.1 | NP_997691.1 | 2 | 99 | ||
| XM_008767283.1 | XP_008765505.1 | 2 | 364 | |||
| GenBank mRNA | AF512994.1 | - | 2 | 99 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcne4| Gene Symbol: Kcne4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212526.1 | NP_997691.1 | 2 | 99 | ||
| XM_008767283.1 | XP_008765505.1 | 2 | 364 | |||
| GenBank mRNA | AF512994.1 | - | 2 | 99 | ||