siRNA Details
| Gene Symbol: Col11a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212528.1 | NP_997693.1 | 5 | 895 | ||
| XM_006255928.2 | XP_006255990.1 | 5 | 1743 | |||
| XM_006255929.2 | XP_006255991.1 | 5 | 1743 | |||
| XM_008772720.1 | XP_008770942.1 | 5 | 1744 | |||
| XM_008772721.1 | XP_008770943.1 | 5 | 1849 | |||
| Gene Symbol: LOC100911886 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008772788.1 | XP_008771010.1 | 5 | 623 | ||

Col11a2
Genomic Map