siRNA Details
| Gene Symbol: Mrps18b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212534.2 | NP_997699.2 | 2 | 145 | ||
| GenBank mRNA | BC166875.1 | - | 2 | 127 | 6889975 | |
| FM036709.1 | - | 2 | 145 | |||
| FQ215273.1 | - | 2 | 118 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mrps18b| Gene Symbol: Mrps18b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212534.2 | NP_997699.2 | 2 | 145 | ||
| GenBank mRNA | BC166875.1 | - | 2 | 127 | 6889975 | |
| FM036709.1 | - | 2 | 145 | |||
| FQ215273.1 | - | 2 | 118 | |||