siRNA Details
| Gene Symbol: SPEN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015001.2 | NP_055816.2 | 2 | 521 | ||
| GenBank mRNA | AF356524.1 | - | 2 | 521 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_019763, AF156529, AB055980, AK137647.

SPEN
Genomic Map