siRNA Details
| Gene Symbol: Prr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212544.2 | NP_997709.1 | 4 | 1035 | ||
| XM_006255942.1 | XP_006256004.1 | 5 | 1103 | |||
| XM_006255943.2 | XP_006256005.1 | 4 | 921 | |||
| GenBank mRNA | BC098636.1 | - | 5 | 1113 | ||
| BC126073.1 | - | 4 | 1035 | 7311139 | ||

Prr3
Genomic Map