siRNA Details
| Gene Symbol: TMEM215 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212558.2 | NP_997723.2 | 2 | 1066 | ||
| GenBank mRNA | AK125320.1 | - | 2 | 1066 | ||
| BX647550.1 | - | 2 | 1025 | |||
| CR749589.1 | - | 1 | 887 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM215| Gene Symbol: TMEM215 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212558.2 | NP_997723.2 | 2 | 1066 | ||
| GenBank mRNA | AK125320.1 | - | 2 | 1066 | ||
| BX647550.1 | - | 2 | 1025 | |||
| CR749589.1 | - | 1 | 887 | |||