siRNA Details
| Gene Symbol: Vars2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_213563.1 | NP_998728.1 | 29 | 4075 | ||
| XM_006255932.1 | XP_006255994.1 | 29 | 4113 | |||
| XM_006255933.1 | XP_006255995.1 | 29 | 3658 | |||
| XM_006255934.1 | XP_006255996.1 | 29 | 4149 | |||
| GenBank mRNA | FQ219325.1 | - | 3 | 1006 | ||

Vars2
Genomic Map