siRNA Details
| Gene Symbol: Zfp57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_213565.4 | NP_998730.2 | 5 | 470 | ||
| XM_008772694.1 | XP_008770916.1 | 4 | 643 | |||
| GenBank mRNA | BC127468.1 | - | 3 | 288 | 8368087 | |
| CA511208.1 | - | 5 | 471 | |||
| DY312591.1 | - | 5 | 522 | |||

Zfp57
Genomic Map