siRNA Details
| Gene Symbol: ZNF517 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134284.1 | 5, 6 | 1685 | |||
| GenBank mRNA | AK131440.1 | - | 5, 6 | 1685 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF517| Gene Symbol: ZNF517 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134284.1 | 5, 6 | 1685 | |||
| GenBank mRNA | AK131440.1 | - | 5, 6 | 1685 | ||