siRNA Details
| Gene Symbol: ZNF517 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317936.1 | NP_001304865.1 | 3 | 189 | ||
| NM_213605.2 | NP_998770.2 | 3 | 189 | |||
| NR_134284.1 | 3 | 189 | ||||
| XM_011517014.2 | XP_011515316.1 | 3 | 728 | |||
| XM_011517015.2 | XP_011515317.1 | 3 | 170 | |||
| GenBank mRNA | AK096527.1 | - | 3 | 728 | ||
| AK097278.1 | - | 2 | 855 | |||
| AK131440.1 | - | 3 | 189 | |||
| BC126161.1 | - | 2 | 109 | 8991877 | ||
| DB016003.1 | - | 3 | 189 | |||
| DB165783.1 | - | 3 | 148 | |||

ZNF517
Genomic Map