siRNA Details
| Gene Symbol: ZNF517 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134284.1 | 6 | 1822 | |||
| XM_011517014.2 | XP_011515316.1 | 5 | 3092 | |||
| XM_011517015.2 | XP_011515317.1 | 5 | 2534 | |||
| XM_011517017.2 | XP_011515319.1 | 4 | 2951 | |||
| XM_017013384.1 | XP_016868873.1 | 2 | 2477 | |||
| GenBank mRNA | AK131440.1 | - | 6 | 1822 | ||

ZNF517
Genomic Map