siRNA Details
| Gene Symbol: FAM19A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252216.1 | NP_001239145.1 | 5 | 1226 | ||
| NM_213609.3 | NP_998774.2 | 5 | 1500 | |||
| XM_017006386.1 | XP_016861875.1 | 4 | 1647 | |||
| XM_017006387.1 | XP_016861876.1 | 5 | 2139 | |||
| XM_017006388.1 | XP_016861877.1 | 5 | 1932 | |||
| XM_017006389.1 | XP_016861878.1 | 4 | 1644 | |||
| XM_017006390.1 | XP_016861879.1 | 5 | 2140 | |||
| GenBank mRNA | AL713702.1 | - | 4 | 1409 | ||
| BC025746.1 | - | 5 | 1402 | 5200142 | ||

FAM19A1
Genomic Map