siRNA Details
| Gene Symbol: RGD1302996 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_213610.3 | NP_998775.2 | 5 | 851 | ||
| XM_006255918.2 | XP_006255980.1 | 5 | 869 | |||
| XM_008772712.1 | XP_008770934.1 | 5 | 740 | |||
| XM_008772713.1 | XP_008770935.1 | 5 | 728 | |||
| GenBank mRNA | BC088194.1 | - | 5 | 851 | 7373688 | |
| FQ215898.1 | - | 5 | 857 | |||
| FQ225840.1 | - | 4 | 634 | |||

RGD1302996
Genomic Map