siRNA Details
| Gene Symbol: Sept5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_213614.2 | NP_998779.2 | 12 | 1407 | ||
| GenBank mRNA | AF033350.1 | - | 10 | 1254 | ||
| AK134700.1 | - | 12 | 1344 | |||
| BC059848.1 | - | 10 | 1285 | |||
| BC079631.1 | - | 6 | 721 | |||
| BC094347.1 | - | 11 | 1283 | |||

Sept5
Genomic Map