siRNA Details
| Gene Symbol: Dnajc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_213625.1 | NP_998790.1 | 7 | 692 | ||
| XM_006247311.2 | XP_006247373.1 | 8 | 1053 | |||
| XM_006247312.2 | XP_006247374.1 | 7 | 773 | |||
| XM_006247313.1 | XP_006247375.1 | 6 | 613 | |||
| GenBank mRNA | AB178470.1 | - | 7 | 692 | ||
| BC093600.1 | - | 6 | 608 | 7316009 | ||
| FQ212948.1 | - | 7 | 723 | |||

Dnajc7
Genomic Map