siRNA Details
| Gene Symbol: Zdhhc23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_213627.2 | NP_998792.2 | 4 | 1543 | ||
| XM_008768738.1 | XP_008766960.1 | 4 | 1641 | |||
| XM_008768739.1 | XP_008766961.1 | 4 | 1563 | |||
| XM_008768740.1 | XP_008766962.1 | 5 | 1805 | |||
| XM_008768741.1 | XP_008766963.1 | 4 | 1554 | |||
| XM_008768742.1 | XP_008766964.1 | 4 | 1452 | |||
| GenBank mRNA | AB098078.1 | - | 5 | 1581 | ||
| AY886538.1 | - | 4 | 1543 | |||

Zdhhc23
Genomic Map