siRNA Details
| Gene Symbol: St8sia5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_213628.1 | NP_998793.1 | 1, 2 | 139 | ||
| GenBank mRNA | AJ699422.1 | - | 1, 2 | 139 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

St8sia5| Gene Symbol: St8sia5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_213628.1 | NP_998793.1 | 1, 2 | 139 | ||
| GenBank mRNA | AJ699422.1 | - | 1, 2 | 139 | ||