siRNA Details
| Gene Symbol: Btla | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_213630.1 | NP_998795.1 | 4 | 510 | ||
| GenBank mRNA | AY590499.1 | - | 4 | 510 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Btla| Gene Symbol: Btla | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_213630.1 | NP_998795.1 | 4 | 510 | ||
| GenBank mRNA | AY590499.1 | - | 4 | 510 | ||