siRNA Details
| Gene Symbol: Amer3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_213727.2 | NP_998892.1 | 2 | 1517 | ||
| GenBank mRNA | AK034979.1 | - | 1 | 1389 | ||
| BC070435.1 | - | 2 | 1483 | 6853754 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Amer3| Gene Symbol: Amer3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_213727.2 | NP_998892.1 | 2 | 1517 | ||
| GenBank mRNA | AK034979.1 | - | 1 | 1389 | ||
| BC070435.1 | - | 2 | 1483 | 6853754 | ||