siRNA Details
Gene Symbol: Krt39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_213730.2 | NP_998895.2 | 7 | 1438 | ||
GenBank mRNA | BC128563.1 | - | 7 | 1433 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Krt39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_213730.2 | NP_998895.2 | 7 | 1438 | ||
GenBank mRNA | BC128563.1 | - | 7 | 1433 |