siRNA Details
| Gene Symbol: PRR27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_214711.3 | NP_999876.2 | 5 | 1273 | ||
| GenBank mRNA | BX640924.1 | - | 5 | 1224 | ||
| BX640997.1 | - | 5 | 1260 | |||
| BX647183.1 | - | 6 | 1097 | |||
| BX647185.1 | - | 7 | 1203 | |||
| BX648724.1 | - | 5 | 1222 | |||
| BX648872.1 | - | 5 | 1146 | |||
| BX648895.1 | - | 5 | 1222 | |||
| BX648896.1 | - | 6 | 1157 | |||

PRR27
Genomic Map