siRNA Details
| Gene Symbol: Olr1386 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_214834.1 | NP_999999.1 | 1 | 551 | ||
| XM_006246176.2 | XP_006246238.1 | 2 | 606 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr1386| Gene Symbol: Olr1386 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_214834.1 | NP_999999.1 | 1 | 551 | ||
| XM_006246176.2 | XP_006246238.1 | 2 | 606 | |||