siRNA Details
| Gene Symbol: GPR137C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099652.1 | NP_001093122.1 | 7 | 1458 | ||
| GenBank mRNA | AK123953.1 | - | 7 | 1354 | ||
| AK293812.1 | - | 5 | 907 | |||
| AL834372.1 | - | 8 | 1366 | |||
| BX647179.1 | - | 5 | 2979 | |||

GPR137C
Genomic Map