siRNA Details
| Gene Symbol: Sirt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008368.1 | NP_001008369.1 | 15 | 1568 | ||
| XM_006228672.2 | XP_006228734.1 | 14 | 1519 | |||
| XM_006228673.2 | XP_006228735.1 | 15 | 1524 | |||
| XM_006228675.1 | XP_006228737.1 | 15 | 1303 | |||
| XM_006228676.2 | XP_006228738.1 | 16 | 1308 | |||
| GenBank mRNA | BC086545.1 | - | 15 | 1568 | 7134377 | |
| FQ212805.1 | - | 15 | 1230 | |||
| FQ216424.1 | - | 11 | 899 | |||

Sirt2
Genomic Map