siRNA Details
| Gene Symbol: Slc25a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057102.1 | NP_476443.1 | 2 | 199 | ||
| GenBank mRNA | BC059108.1 | - | 2 | 248 | 6922359 | |
| D12771.1 | - | 2 | 199 | |||
| FQ213290.1 | - | 2 | 221 | |||
| FQ213647.1 | - | 2 | 221 | |||
| FQ213855.1 | - | 2 | 239 | |||
| FQ214507.1 | - | 2 | 176 | |||
| FQ219677.1 | - | 2 | 221 | |||
| FQ220249.1 | - | 2 | 223 | |||
| FQ220272.1 | - | 2 | 223 | |||
| FQ220435.1 | - | 2 | 243 | |||
| FQ229325.1 | - | 2 | 224 | |||
| FQ229438.1 | - | 2 | 224 | |||

Slc25a5
Genomic Map