siRNA Details
| Gene Symbol: Clmp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173154.1 | NP_775177.1 | 7 | 1849 | ||
| GenBank mRNA | AF302047.1 | - | 7 | 1849 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Clmp| Gene Symbol: Clmp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173154.1 | NP_775177.1 | 7 | 1849 | ||
| GenBank mRNA | AF302047.1 | - | 7 | 1849 | ||