siRNA Details
| Gene Symbol: EXPH5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001308019.1 | NP_001294948.1 | 2 | 61 | ||
| NM_015065.2 | NP_055880.2 | 1 | 188 | |||
| XM_011542696.2 | XP_011540998.1 | 1 | 14 | |||
| XM_017017397.1 | XP_016872886.1 | 2 | 77 | |||
| XM_017017398.1 | XP_016872887.1 | 2 | 77 | |||
| GenBank mRNA | AB014524.1 | - | 2 | 61 | ||
| AY099469.1 | - | 1 | 174 | |||
| BC113119.1 | - | 1 | 159 | 40037748 | ||
| BC117701.1 | - | 1 | 159 | 40074153 | ||
| BP358360.1 | - | 1 | 188 | |||

EXPH5
Genomic Map