siRNA Details
| Gene Symbol: Tpte | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199257.2 | NP_954866.2 | 28 | 2296 | ||
| GenBank mRNA | AJ311311.1 | - | 28 | 2299 | ||
| AJ311312.1 | - | 28 | 2384 | |||
| AJ311313.1 | - | 29 | 2422 | |||
| BC107329.2 | - | 28 | 2091 | 40054415 | ||

Tpte
Genomic Map