siRNA Details
Gene Symbol: Amigo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182816.2 | NP_877968.2 | 1 | 2578 | ||
XM_006242290.1 | XP_006242352.1 | 1 | 1942 | |||
GenBank mRNA | AB078879.1 | - | 1 | 2578 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Amigo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182816.2 | NP_877968.2 | 1 | 2578 | ||
XM_006242290.1 | XP_006242352.1 | 1 | 1942 | |||
GenBank mRNA | AB078879.1 | - | 1 | 2578 |