siRNA Details
| Gene Symbol: Strada | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182820.1 | NP_877972.1 | 10 | 1146 | ||
| XM_006247586.2 | XP_006247648.1 | 12 | 1325 | |||
| XM_006247588.2 | XP_006247650.1 | 12 | 1324 | |||
| XM_006247595.2 | XP_006247657.1 | 11 | 1294 | |||
| XM_008768359.1 | XP_008766581.1 | 12 | 1325 | |||
| XM_008768360.1 | XP_008766582.1 | 12 | 1301 | |||
| XM_008768361.1 | XP_008766583.1 | 10 | 1237 | |||
| XM_008768362.1 | XP_008766584.1 | 10 | 1213 | |||
| XM_008768363.1 | XP_008766585.1 | 11 | 1295 | |||
| XM_008768364.1 | XP_008766586.1 | 11 | 1271 | |||
| GenBank mRNA | AY327409.1 | - | 10 | 1146 | ||
| BC081911.1 | - | 11 | 1230 | 7111907 | ||

Strada
Genomic Map