siRNA Details
| Gene Symbol: Arhgef4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183019.2 | NP_898840.2 | 10 | 1817 | ||
| GenBank mRNA | AK034013.1 | - | 10 | 1781 | ||
| AK122448.2 | - | 6 | 872 | |||
| AK134629.1 | - | 1780 | ||||
| AK138598.1 | - | 10 | 1777 | |||
| AK163424.1 | - | 10 | 1799 | |||
| AK169957.1 | - | 10 | 1837 | |||
| BC055804.1 | - | 10 | 1778 | |||
| BC094608.1 | - | 5337 | ||||
| BC139371.1 | - | 10 | 1800 | 8862393 | ||
| BC139397.1 | - | 10 | 1800 | 8862419 | ||

Arhgef4
Genomic Map