siRNA Details
| Gene Symbol: Fam19a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183224.3 | NP_899047.1 | 7 | 4111 | ||
| GenBank mRNA | AK078681.1 | - | 7 | 4114 | ||
| AV360034.2 | - | 1 | 478 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam19a3| Gene Symbol: Fam19a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183224.3 | NP_899047.1 | 7 | 4111 | ||
| GenBank mRNA | AK078681.1 | - | 7 | 4114 | ||
| AV360034.2 | - | 1 | 478 | |||