siRNA Details
| Gene Symbol: Srpk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_184045.2 | NP_908934.1 | 8, 9 | 805 | ||
| GenBank mRNA | AY346151.1 | - | 8, 9 | 809 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Srpk3| Gene Symbol: Srpk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_184045.2 | NP_908934.1 | 8, 9 | 805 | ||
| GenBank mRNA | AY346151.1 | - | 8, 9 | 809 | ||