siRNA Details
| Gene Symbol: Onecut2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_194268.2 | NP_919244.2 | 2 | 1537 | ||
| GenBank mRNA | AY242995.1 | - | 2 | 1537 | ||
| BC100750.1 | - | 3 | 1034 | |||
| BC100755.1 | - | 3 | 1034 | |||
| BC103668.1 | - | 3 | 1370 | 40046651 | ||

Onecut2
Genomic Map